Ethyl 1-benzoylpiperidine-4-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: 1-Benzoyl-4-(ethoxycarbonyl)piperidine, Phenyl[4-(ethoxycarbonyl)piperidin-1-yl]methanone

OR9920-10G OR9920-25G
APOSOR9920-10GEA 87.3 EUR
APOSOR9920-10G APOSOR9920-25G
Ethyl 1-benzoylpiperidine-4-carboxylate 97%
Ethyl 1-benzoylpiperidine-4-carboxylate
Formel: C₁₅H₁₉NO₃
Siedepunkt: 169…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00667804
CAS-Nummer: 136081-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR