4-Chloro-2-(chloromethyl)-3-methoxypyridine hydrochloride, 90%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Chloro-3-methoxypicolinyl chloride hydrochloride

OR9946-25G OR9946-5G OR9946-100G
APOSOR9946-25GEA 633 EUR
APOSOR9946-25G APOSOR9946-5G APOSOR9946-100G
4-Chloro-2-(chloromethyl)-3-methoxypyridine hydrochloride, 90%
4-Chloro-2-(chloromethyl)-3-methoxypyridine hydrochloride,
Formel: C₇H₇Cl₂NO·ClH
Molecular Weight: 228.504 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07369084
CAS-Nummer: 503058-51-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR