3-Bromo-4-phenylpyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Bromopyridin-4-yl)benzene

OR9978-1G OR9978-250MG OR9978-5G
APOSOR9978-1GEA 132 EUR
APOSOR9978-1G APOSOR9978-250MG APOSOR9978-5G
3-Bromo-4-phenylpyridine
3-Bromo-4-phenylpyridine
Formel: C₁₁H₈BrN
Molecular Weight: 234,092 g/mol
Schmelzpunkt: 46…48 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114247
CAS-Nummer: 88345-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR