4-Chloro-2,6-difluorobenzyl alcohol 98%

Lieferant: Apollo Scientific

PC0004-10G PC0004-5G
APOSPC0004-10GEA 260 EUR
APOSPC0004-10G APOSPC0004-5G
4-Chloro-2,6-difluorobenzyl alcohol 98%
4-Chloro-2,6-difluorobenzyl alcohol
Formel: C₇H₅ClF₂O
Molecular Weight: 178,564 g/mol
Schmelzpunkt: 73…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095327
CAS-Nummer: 252004-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR