5-(4-Bromophenyl)-2-(trifluoromethyl)-3-furoic acid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-(4-Bromophenyl)-2-(trifluoromethyl)furan-3-carboxylic acid, 5-(4-Bromophenyl)-3-carboxy-2-(trifluoromethyl)furan, 1-Bromo-4-[4-carboxy-5-(trifluoromethyl)fur-2-yl]benzene
PC0059-1G
PC0059-5G
APOSPC0059-1GEA
104
EUR
APOSPC0059-1G
APOSPC0059-5G
5-(4-Bromophenyl)-2-(trifluoromethyl)-3-furoic acid
5-(4-Bromophenyl)-2-(trifluoromethyl)-3-furoic acid
Formel:
C₁₂H₆BrF₃O₃ Schmelzpunkt: 199…202 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01313498 CAS-Nummer: 229957-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...