6-(Perfluorohexyl)hexanol 96%

Lieferant: Apollo Scientific

PC0078-1G PC0078-5G
APOSPC0078-1GEA 212 EUR
APOSPC0078-1G APOSPC0078-5G
6-(Perfluorohexyl)hexanol 96%
6-(Perfluorohexyl)hexanol
Formel: C₁₂H₁₃F₁₃O
Molecular Weight: 420.21 g/mol
Dichte: 1.493 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00153241
CAS-Nummer: 161981-35-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR