3-Chlor-4-fluorzimtsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(3-Chlor-4-fluorphenyl)propensäure, 3-Chlor-4-fluorcinnamsäure

PC0084-1G PC0084-25G PC0084-5G
APOSPC0084-1GEA 21.9 EUR
APOSPC0084-1G APOSPC0084-25G APOSPC0084-5G
3-Chlor-4-fluorzimtsäure 95%
3-Chlor-4-fluorzimtsäure
Formel: C₉H₆ClFO₂
Molecular Weight: 200.6 g/mol
Schmelzpunkt: 173…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631393
CAS-Nummer: 58537-11-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR