2-Chloro-6-fluoro-3-methylbenzyl bromide

Lieferant: Apollo Scientific
Gefahr

PC0114-10G PC0114-5G PC0114-1G
APOSPC0114-10GEA 314 EUR
APOSPC0114-10G APOSPC0114-5G APOSPC0114-1G
2-Chloro-6-fluoro-3-methylbenzyl bromide
2-Chloro-6-fluoro-3-methylbenzyl bromide
Formel: C₈H₇BrClF
Molecular Weight: 237,497 g/mol
Schmelzpunkt: 35…37 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631363
CAS-Nummer: 261762-87-2
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR