Bis(2,2,2-trifluoroethyl)maleate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Bis(2,2,2-trifluoroethyl) (2Z)-but-2-ene-1,4-dioate, Maleic acid bis(2,2,2-trifluoroethyl)ester
PC0148-25G
PC0148-5G
APOSPC0148-25GEA
190
EUR
APOSPC0148-25G
APOSPC0148-5G
Bis(2,2,2-trifluoroethyl)maleate 97%
Bis(2,2,2-trifluoroethyl)maleate
Formel:
C₈H₆F₆O₄ Molecular Weight: 280,121 g/mol Siedepunkt: 50 °C (144 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078313 CAS-Nummer: 116401-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...