4-Amino-2-fluoro-3-(trifluoromethyl)benzonitrile 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Cyano-3-fluoro-2-(trifluoromethyl)aniline, 6-Amino-3-cyano-2-fluorobenzotrifluoride

PC01486-1G PC01486-250MG
APOSPC01486-1GEA 575 EUR
APOSPC01486-1G APOSPC01486-250MG
4-Amino-2-fluoro-3-(trifluoromethyl)benzonitrile 95%
4-Amino-2-fluoro-3-(trifluoromethyl)benzonitrile
Formel: C₈H₄F₄N₂
Molecular Weight: 204,124 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25976371
CAS-Nummer: 1448858-64-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR