2'-Amino-2,2,2,3'-tetrafluoroacetophenone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-6-(trifluoroacetyl)aniline, 1-(2-Amino-3-fluorophenyl)-2,2,2-trifluoroethan-1-one, 1-(2-Amino-3-fluorophenyl)-2,2,2-trifluoroethanone
PC01504-1G
PC01504-250MG
APOSPC01504-1GEA
1110
EUR
APOSPC01504-1G
APOSPC01504-250MG
2'-Amino-2,2,2,3'-tetrafluoroacetophenone
2'-Amino-2,2,2,3'-tetrafluoroacetophenone
Formel:
C₈H₅F₄NO Molecular Weight: 207,125 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25976372 CAS-Nummer: 1448858-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...