2-Amino-5-iodo-4-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Iodo-4-(trifluoromethyl)pyridin-2-amine

PC01522-1G PC01522-250MG
APOSPC01522-1GEA 400 EUR
APOSPC01522-1G APOSPC01522-250MG
2-Amino-5-iodo-4-(trifluoromethyl)pyridine
2-Amino-5-iodo-4-(trifluoromethyl)pyridine
Formel: C₆H₄F₃IN₂
Molecular Weight: 288,009 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22123874
CAS-Nummer: 1321530-57-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR