4-(Methylsulphonyl)-2-(trifluoromethyl)benzaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Formyl-3-(trifluoromethyl)phenyl methyl sulphone, 2-Formyl-5-(methylsulphonyl)benzotrifluoride

PC01643-1G PC01643-5G
APOSPC01643-1GEA 21.9 EUR
APOSPC01643-1G APOSPC01643-5G
4-(Methylsulphonyl)-2-(trifluoromethyl)benzaldehyde
4-(Methylsulphonyl)-2-(trifluoromethyl)benzaldehyde
Formel: C₉H₇F₃O₃S
Molecular Weight: 252,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15479075
CAS-Nummer: 1215310-75-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR