4-(Methylsulphonyl)-2-(trifluoromethyl)benzyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(Chloromethyl)-3-(trifluoromethyl)phenyl methyl sulphone, 2-(Chloromethyl)-5-(methylsulphonyl)benzotrifluoride

PC01644-1G PC01644-5G
APOSPC01644-1GEA 120 EUR
APOSPC01644-1G APOSPC01644-5G
4-(Methylsulphonyl)-2-(trifluoromethyl)benzyl chloride
4-(Methylsulphonyl)-2-(trifluoromethyl)benzyl chloride
Formel: C₉H₈ClF₃O₂S
Molecular Weight: 272,672 g/mol
Schmelzpunkt: 91…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11046211
CAS-Nummer: 1086389-83-4
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR