2-Fluoro-4-methoxy-α-methylbenzyl alcohol 97%
Lieferant: Apollo Scientific
Synonyme:
1-(2-Fluoro-4-methoxyphenyl)ethan-1-ol, 1-(2-Fluoro-4-methoxyphenyl)ethanol
PC01648-1G
PC01648-250MG
PC01648-5G
APOSPC01648-1GEA
290
EUR
APOSPC01648-1G
APOSPC01648-250MG
APOSPC01648-5G
2-Fluoro-4-methoxy-α-methylbenzyl alcohol 97%
2-Fluoro-4-methoxy-α-methylbenzyl alcohol
Formel:
C₉H₁₁FO₂ Molecular Weight: 170,181 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12047707 CAS-Nummer: 74457-87-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...