2'-Amino-3'-chloro-2,2,2-trifluoroacetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-6-(trifluoroacetyl)aniline, 1-(2-Amino-3-chlorophenyl)-2,2,2-trifluoroethan-1-one, 1-(2-Amino-3-chlorophenyl)-2,2,2-trifluoroethanone

PC01673-100MG PC01673-1G PC01673-250MG
APOSPC01673-100MGEA 154 EUR
APOSPC01673-100MG APOSPC01673-1G APOSPC01673-250MG
2'-Amino-3'-chloro-2,2,2-trifluoroacetophenone
2'-Amino-3'-chloro-2,2,2-trifluoroacetophenone
Formel: C₈H₅ClF₃NO
Molecular Weight: 223,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22689228
CAS-Nummer: 1414958-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR