2'-Amino-2,2,2-trifluoro-4'-(trifluoromethyl)acetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Amino-4-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone, 3-Amino-4-(trifluoroacetyl)benzotrifluoride, 1-[2-Amino-4-(trifluoromethyl)phenyl]-2,2,2-trifluoroethan-1-one, 2-(Trifluoroacetyl)-5-(trifluoromethyl)aniline

PC01674-1G PC01674-250MG
APOSPC01674-1GEA 395 EUR
APOSPC01674-1G APOSPC01674-250MG
2'-Amino-2,2,2-trifluoro-4'-(trifluoromethyl)acetophenone
2'-Amino-2,2,2-trifluoro-4'-(trifluoromethyl)acetophenone
Formel: C₉H₅F₆NO
Molecular Weight: 257.133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25976379
CAS-Nummer: 1448858-61-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR