2-(Difluoromethyl)benzene boronic acid, pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-(Difluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-[2-(Difluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PC01681-1G PC01681-250MG
APOSPC01681-1GEA 406 EUR
APOSPC01681-1G APOSPC01681-250MG
2-(Difluoromethyl)benzene boronic acid, pinacol ester
2-(Difluoromethyl)benzene boronic acid, pinacol ester
Formel: C₁₃H₁₇BF₂O₂
Molecular Weight: 254,081 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16996279
CAS-Nummer: 879275-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR