3-Chloro-4-(trifluoromethoxy)benzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: [3-Chloro-4-(trifluoromethoxy)phenyl]methanol, (3-Chloro-4-(trifluoromethoxy)phenyl)methanol

PC0180-5G PC0180-1G
APOSPC0180-5GEA 154 EUR
APOSPC0180-5G APOSPC0180-1G
3-Chloro-4-(trifluoromethoxy)benzyl alcohol 97%
3-Chloro-4-(trifluoromethoxy)benzyl alcohol
Formel: C₈H₆ClF₃O₂
Molecular Weight: 226,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631559
CAS-Nummer: 56456-48-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR