4-Bromo-5-phenyl-3-(trifluoromethyl)-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
[4-Bromo-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzene
PC0223-1G
PC0223-25G
PC0223-5G
APOSPC0223-1GEA
29.2
EUR
APOSPC0223-1G
APOSPC0223-25G
APOSPC0223-5G
4-Bromo-5-phenyl-3-(trifluoromethyl)-1H-pyrazole
4-Bromo-5-phenyl-3-(trifluoromethyl)-1H-pyrazole
Formel:
C₁₀H₆BrF₃N₂ Schmelzpunkt: 173…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153731 CAS-Nummer: 230295-07-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...