2,6-Difluoro-3-methylbenzyl alcohol 98%

Lieferant: Apollo Scientific

Synonyme: (2,6-Difluoro-3-methylphenyl)methanol, 2,4-Difluoro-3-(hydroxymethyl)toluene

PC0250-10G PC0250-5G
APOSPC0250-10GEA 197 EUR
APOSPC0250-10G APOSPC0250-5G
2,6-Difluoro-3-methylbenzyl alcohol 98%
2,6-Difluoro-3-methylbenzyl alcohol
Formel: C₈H₈F₂O
Molecular Weight: 158,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631335
CAS-Nummer: 261763-40-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR