2,5-Bis(trifluoromethyl)benzeneboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5-Bis(trifluoromethyl)phenylboronic acid

PC0453-1G PC0453-5G
APOSPC0453-1GEA 69.6 EUR
APOSPC0453-1G APOSPC0453-5G
2,5-Bis(trifluoromethyl)benzeneboronic acid
2,5-Bis(trifluoromethyl)benzeneboronic acid
Formel: C₈H₅BF₆O₂
Molecular Weight: 257.926 g/mol
Schmelzpunkt: 133…135 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04038223
CAS-Nummer: 196083-18-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR