2,3,4-Trifluoro-6-(trifluoromethyl)benzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC0534-1G
PC0534-250MG
APOSPC0534-1GEA
870
EUR
APOSPC0534-1G
APOSPC0534-250MG
2,3,4-Trifluoro-6-(trifluoromethyl)benzoic acid
2,3,4-Trifluoro-6-(trifluoromethyl)benzoic acid
Formel:
C₈H₂F₆O₂ Molecular Weight: 244,091 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631432 CAS-Nummer: 157337-84-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...