(1R)-1-(2-Fluorophenyl)ethan-1-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-2-Fluoro-α-methylbenzyl alcohol, (R)-1-(2-Fluorophenyl)ethanol

PC0603-1G PC0603-25G PC0603-5G
APOSPC0603-1GEA 34.8 EUR
APOSPC0603-1G APOSPC0603-25G APOSPC0603-5G
(1R)-1-(2-Fluorophenyl)ethan-1-ol
(1R)-1-(2-Fluorophenyl)ethan-1-ol
Formel: C₈H₉FO
Molecular Weight: 140,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03092991
CAS-Nummer: 162427-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR