(S)-1-(3-Fluorophenyl)ethanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
S-MF-PEL
PC0608-1G
PC0608-250MG
PC0608-5G
APOSPC0608-1GEA
212
EUR
APOSPC0608-1G
APOSPC0608-250MG
APOSPC0608-5G
(S)-1-(3-Fluorophenyl)ethanol 97%
(S)-1-(3-Fluorophenyl)ethanol
Formel:
C₈H₉FO Molecular Weight: 140,155 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03092996 CAS-Nummer: 126534-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...