(1R)-1-(2-Fluorophenyl)ethylamine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(R)-1-(2-Fluorophenyl)ethanamine
PC0611-100MG
PC0611-250MG
APOSPC0611-100MGEA
33.5
EUR
APOSPC0611-100MG
APOSPC0611-250MG
(1R)-1-(2-Fluorophenyl)ethylamine
(1R)-1-(2-Fluorophenyl)ethylamine
Formel:
C₈H₁₀FN Molecular Weight: 139,17 g/mol Dichte: 1 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03092993 CAS-Nummer: 185545-90-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...