(3S)-3-Hydroxy-4,4,4-trifluorobutanoic acid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(3S)-3-Hydroxy-4,4,4-trifluorobutyric acid
PC0622-1G
PC0622-250MG
PC0622-100MG
APOSPC0622-1GEA
1940
EUR
APOSPC0622-1G
APOSPC0622-250MG
APOSPC0622-100MG
(3S)-3-Hydroxy-4,4,4-trifluorobutanoic acid 97%
(3S)-3-Hydroxy-4,4,4-trifluorobutanoic acid
Formel:
C₄H₅F₃O₃ Molecular Weight: 158,076 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03093018 CAS-Nummer: 128899-79-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...