Hexakis(1H,1H-perfluoropropoxy)phosphazene 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC0725-1G
PC0725-250MG
APOSPC0725-1GEA
475
EUR
APOSPC0725-1G
APOSPC0725-250MG
Hexakis(1H,1H-perfluoropropoxy)phosphazene 95%
Hexakis(1H,1H-perfluoropropoxy)phosphazene
Formel:
C₁₈H₁₂F₃₀N₃O₆P₃ Molecular Weight: 1029,178 g/mol Siedepunkt: 120…122 °C (2,5 mm Hg) Schmelzpunkt: 14…16 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798141 CAS-Nummer: 429-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...