3-(Trifluoromethyl)dihydrofuran-2(3H)-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
α-(Trifluoromethyl)-γ-butyrolactone, 2-Oxo-3-(trifluoromethyl)tetrahydrofuran, 3-(Trifluoromethyl)oxolan-2-one
PC0793-250MG
PC0793-100MG
APOSPC0793-250MGEA
111
EUR
APOSPC0793-250MG
APOSPC0793-100MG
3-(Trifluoromethyl)dihydrofuran-2(3H)-one
3-(Trifluoromethyl)dihydrofuran-2(3H)-one
Formel:
C₅H₅F₃O₂ Molecular Weight: 154,087 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798462 CAS-Nummer: 174744-18-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...