1-(4-Fluorophenyl)-5-phenylpenta-2,4-dien-1-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Fluorobenzoyl)-4-phenylbuta-1,3-diene

PC0821-100MG PC0821-1G
APOSPC0821-100MGEA 34.8 EUR
APOSPC0821-100MG APOSPC0821-1G
1-(4-Fluorophenyl)-5-phenylpenta-2,4-dien-1-one
1-(4-Fluorophenyl)-5-phenylpenta-2,4-dien-1-one
Formel: C₁₇H₁₃FO
Molecular Weight: 252,283 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017966
CAS-Nummer: 99646-84-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR