Hexakis(2,2,3,3-tetrafluoropropoxy)phosphazene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,2,4,4,6,6-Hexakis(2,2,3,3-tetrafluoropropoxy)-1,3,5-triazatriphosphine, Hexakis(1H,1H,3H-perfluoropropoxy)phosphazene, Hexakis(1H,1H,3H-tetrafluoropropoxy)phosphazene
PC0874-1G
PC0874-250MG
APOSPC0874-1GEA
725
EUR
APOSPC0874-1G
APOSPC0874-250MG
Hexakis(2,2,3,3-tetrafluoropropoxy)phosphazene
Hexakis(2,2,3,3-tetrafluoropropoxy)phosphazene
Formel:
C₁₈H₁₈F₂₄N₃O₆P₃ Molecular Weight: 921,235 g/mol Siedepunkt: 174…176 °C (2 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798140 CAS-Nummer: 58943-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...