3-Fluoro-2-methylbenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Fluoro-2-methylphenyl)methanol, (3-Fluoro-o-tolyl)methanol

PC0885-10G PC0885-1G PC0885-5G
APOSPC0885-10GEA 109 EUR
APOSPC0885-10G APOSPC0885-1G APOSPC0885-5G
3-Fluoro-2-methylbenzyl alcohol
3-Fluoro-2-methylbenzyl alcohol
Formel: C₈H₉FO
Molecular Weight: 140.155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03788542
CAS-Nummer: 500912-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR