2-Fluoro-6-methoxyphenylacetonitrile 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Cyanomethyl)-3-fluoroanisole, 2-Fluoro-6-methoxybenzyl cyanide, 2-(2-Fluoro-6-methoxyphenyl)acetonitrile
PC0905-1G
PC0905-5G
APOSPC0905-1GEA
84.1
EUR
APOSPC0905-1G
APOSPC0905-5G
2-Fluoro-6-methoxyphenylacetonitrile 95+%
2-Fluoro-6-methoxyphenylacetonitrile
Formel:
C₉H₈FNO Molecular Weight: 165,164 g/mol Siedepunkt: 109…110 °C (3 mm Hg) Lagertemperatur: Kühlschrank |
MDL:
MFCD00671772 CAS-Nummer: 500912-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...