1H,2H-Hexafluorocyclopent-1-ene 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC0970-1G
PC0970-5G
APOSPC0970-1GEA
132
EUR
APOSPC0970-1G
APOSPC0970-5G
1H,2H-Hexafluorocyclopent-1-ene 97%
1H,2H-Hexafluorocyclopent-1-ene
Formel:
C₅H₂F₆ Molecular Weight: 176,06 g/mol Siedepunkt: 70…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03792790 CAS-Nummer: 1005-73-8 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...