(Hydroxymethyl)perfluorocyclohexane

Lieferant: Apollo Scientific
Achtung

Synonyme: (Perfluorocyclohexyl)methanol, (Hydroxymethyl)undecafluorocyclohexane

PC10004-1G
APOSPC10004-1GEA 66.7 EUR
APOSPC10004-1G
(Hydroxymethyl)perfluorocyclohexane
(Hydroxymethyl)perfluorocyclohexane
Formel: C₇H₃F₁₁O
Molecular Weight: 312,081 g/mol
Siedepunkt: 144…146 °C
Dichte: 1,7 g/cm³ (25 °C)
Flammpunkt: 63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00126873
CAS-Nummer: 28788-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR