2-Bromo-6-fluorobenzenesulphonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

PC10329-1G PC10329-5G
APOSPC10329-1GEA 243 EUR
APOSPC10329-1G APOSPC10329-5G
2-Bromo-6-fluorobenzenesulphonyl chloride 95%
2-Bromo-6-fluorobenzenesulphonyl chloride
Formel: C₆H₃BrClFO₂S
Molecular Weight: 273,507 g/mol
Schmelzpunkt: 29…30 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368673
CAS-Nummer: 886762-59-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR