N-[(2,4-Difluorophenyl)carbamothioyl]benzamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Benzoyl-3-(2,4-difluorophenyl)thiourea

PC10450-25G PC10450-5G
APOSPC10450-25GEA 743 EUR
APOSPC10450-25G APOSPC10450-5G
N-[(2,4-Difluorophenyl)carbamothioyl]benzamide
N-[(2,4-Difluorophenyl)carbamothioyl]benzamide
Formel: C₁₄H₁₀F₂N₂OS
Molecular Weight: 292,304 g/mol
Schmelzpunkt: 117…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368702
CAS-Nummer: 887266-92-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR