N-[(2,5-Difluorophenyl)carbamothioyl]benzamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Benzoyl-3-(2,5-difluorophenyl)thiourea, N-((2,5-Difluorophenyl)carbamothioyl)benzamide
PC10451-25G
PC10451-5G
APOSPC10451-25GEA
278
EUR
APOSPC10451-25G
APOSPC10451-5G
N-[(2,5-Difluorophenyl)carbamothioyl]benzamide
N-[(2,5-Difluorophenyl)carbamothioyl]benzamide
Formel:
C₁₄H₁₀F₂N₂OS Molecular Weight: 292,304 g/mol Schmelzpunkt: 166…168,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368703 CAS-Nummer: 887266-93-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...