3-Amino-4,4,4-trifluorobutanamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-4,4,4-trifluorobutyramide
PC10615-1G
PC10615-25G
PC10615-5G
APOSPC10615-1GEA
102
EUR
APOSPC10615-1G
APOSPC10615-25G
APOSPC10615-5G
3-Amino-4,4,4-trifluorobutanamide
3-Amino-4,4,4-trifluorobutanamide
Formel:
C₄H₇F₃N₂O Molecular Weight: 156,106 g/mol Schmelzpunkt: 120…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368716 CAS-Nummer: 453-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...