2-Methyl-4-(trifluoromethoxy)benzoyl chloride 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-(Trifluoromethoxy)-o-toluoyl chloride, 2-(Chlorocarbonyl)-5-(trifluoromethoxy)toluene
PC1063-1G
PC1063-5G
APOSPC1063-1GEA
132
EUR
APOSPC1063-1G
APOSPC1063-5G
2-Methyl-4-(trifluoromethoxy)benzoyl chloride 95+%
2-Methyl-4-(trifluoromethoxy)benzoyl chloride
Formel:
C₉H₆ClF₃O₂ Molecular Weight: 238,591 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631544 CAS-Nummer: 261951-93-3 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...