6-Chloro-5-fluoronicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-3-fluoropyridine-5-carboxylic acid, 6-Chloro-5-fluoropyridine-3-carboxylic acid, 5-Carboxy-2-chloro-3-fluoropyridine

PC10650-1G PC10650-250MG
APOSPC10650-1GEA 50.9 EUR
APOSPC10650-1G APOSPC10650-250MG
6-Chloro-5-fluoronicotinic acid
6-Chloro-5-fluoronicotinic acid
Formel: C₆H₃ClFNO₂
Molecular Weight: 175.545 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226507
CAS-Nummer: 38186-86-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR