2-(Aminomethyl)-5-bromo-3-fluoropyridine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-3-fluoropicolinylamine hydrochloride, (5-Bromo-3-fluoropyridin-2-yl)methanamine hydrochloride, (5-Bromo-3-fluoropyridin-2-yl)methylamine hydrochloride

PC10652-250MG PC10652-500MG
APOSPC10652-250MGEA 69.6 EUR
APOSPC10652-250MG APOSPC10652-500MG
2-(Aminomethyl)-5-bromo-3-fluoropyridine hydrochloride
2-(Aminomethyl)-5-bromo-3-fluoropyridine hydrochloride
Formel: C₆H₆BrFN₂·ClH
Molecular Weight: 241,489 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17214208
CAS-Nummer: 1257535-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR