5-Fluoro-2-methoxynicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Carboxy-5-fluoro-2-methoxypyridine, 5-Fluoro-2-methoxypyridine-3-carboxylic acid

PC10656-1G PC10656-5G
APOSPC10656-1GEA 122 EUR
APOSPC10656-1G APOSPC10656-5G
5-Fluoro-2-methoxynicotinic acid
5-Fluoro-2-methoxynicotinic acid
Formel: C₇H₆FNO₃
Molecular Weight: 171,126 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04972399
CAS-Nummer: 884494-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR