2-Bromo-6-nitro-4-[(trifluoromethyl)sulphonyl]aniline 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-3-bromo-5-[(trifluoromethyl)sulphonyl]nitrobenzene, 4-Amino-3-bromo-5-nitrophenyl trifluoromethyl sulphone

PC10663-1G PC10663-250MG PC10663-5G
APOSPC10663-1GEA 116 EUR
APOSPC10663-1G APOSPC10663-250MG APOSPC10663-5G
2-Bromo-6-nitro-4-[(trifluoromethyl)sulphonyl]aniline 95%
2-Bromo-6-nitro-4-[(trifluoromethyl)sulphonyl]aniline
Formel: C₇H₄BrF₃N₂O₄S
Molecular Weight: 349,082 g/mol
Schmelzpunkt: 147 °C
Lagertemperatur: Kühlschrank
MDL: MFCD22628762
CAS-Nummer: 156425-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR