2-Nitro-4-[(trifluoromethyl)sulphonyl]aniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-[(trifluoromethyl)sulphonyl]nitrobenzene, 4-Amino-3-nitrophenyl trifluoromethyl sulphone

PC10664-1G PC10664-5G
APOSPC10664-1GEA 109 EUR
APOSPC10664-1G APOSPC10664-5G
2-Nitro-4-[(trifluoromethyl)sulphonyl]aniline
2-Nitro-4-[(trifluoromethyl)sulphonyl]aniline
Formel: C₇H₅F₃N₂O₄S
Molecular Weight: 270,186 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00523745
CAS-Nummer: 400-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR