3-Iodo-4-(trifluoromethyl)benzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Carboxy-2-iodobenzotrifluoride, 3-Iodo-α,α,α-trifluoro-p-toluic acid

PC10668-1G PC10668-5G PC10668-25G
APOSPC10668-1GEA 29 EUR
APOSPC10668-1G APOSPC10668-5G APOSPC10668-25G
3-Iodo-4-(trifluoromethyl)benzoic acid
3-Iodo-4-(trifluoromethyl)benzoic acid
Formel: C₈H₄F₃IO₂
Molecular Weight: 316,016 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18398157
CAS-Nummer: 1034690-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR