3,3,3-Trifluoroprop-1-yl toluene-4-sulphonate

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,3,3-Trifluoroprop-1-yl 4-methylbenzenesulphonate, 3,3,3-Trifluoroprop-1-yl tosylate

PC10676-25G PC10676-1G PC10676-5G
APOSPC10676-25GEA 836 EUR
APOSPC10676-25G APOSPC10676-1G APOSPC10676-5G
3,3,3-Trifluoroprop-1-yl toluene-4-sulphonate
3,3,3-Trifluoroprop-1-yl toluene-4-sulphonate
Formel: C₁₀H₁₁F₃O₃S
Molecular Weight: 268,253 g/mol
Siedepunkt: 120…122 °C (1 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD16620595
CAS-Nummer: 2342-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR