2-Fluoro-3-methoxyphenacyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-2'-fluoro-3'-methoxyacetophenone, 2-Bromo-1-(2-fluoro-3-methoxyphenyl)ethan-1-one

PC10678-1G PC10678-250MG
APOSPC10678-1GEA 348 EUR
APOSPC10678-1G APOSPC10678-250MG
2-Fluoro-3-methoxyphenacyl bromide 97%
2-Fluoro-3-methoxyphenacyl bromide
Formel: C₉H₈BrFO₂
Molecular Weight: 247,061 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23711723
CAS-Nummer: 1427363-68-5
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR