2-Iodo-4-(trifluoromethyl)benzyl bromide 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(Bromomethyl)-2-iodo-4-(trifluoromethyl)benzene, 4-(Bromomethyl)-3-iodobenzotrifluoride
PC10681-1G
PC10681-250MG
PC10681-5G
APOSPC10681-1GEA
179
EUR
APOSPC10681-1G
APOSPC10681-250MG
APOSPC10681-5G
2-Iodo-4-(trifluoromethyl)benzyl bromide 97%
2-Iodo-4-(trifluoromethyl)benzyl bromide
Formel:
C₈H₅BrF₃I Molecular Weight: 364,929 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08543940 CAS-Nummer: 939758-31-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...