Fluoromethyl 2-pyridyl sulphone 95%

Lieferant: Apollo Scientific

PC10695-1G
APOSPC10695-1GEA 186 EUR
APOSPC10695-1G
Fluoromethyl 2-pyridyl sulphone 95%
Fluoromethyl 2-pyridyl sulphone
Formel: C₆H₆FNO₂S
Molecular Weight: 175,181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28100935
CAS-Nummer: 1365765-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR